Learn More
asecos™ Dispensing Can
The Dispensing Can allows for safe, clean and comfortable dispensing of liquids in the workplace.
1126.45 NOK - 1438.29 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17816496
|
asecos™
36487 |
2 L |
1126.45 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17836446
|
asecos™
36488 |
5 L |
1260.11 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17856546
|
asecos™
36489 |
10 L |
1438.29 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- Resistant, chemically stable and UV-stable containers
- Incl. 2 screw-on type outlets in diff erent sizes
- 18 labels in 9 signal colours and a multilingual safety label for content labelling according to GHS
- With stable handles for ergnomic handling
Specifications
White | |
Polyethylene |