Learn More
Thermo Scientific™ CTC Instruments Fixed GT Needle Syringe, 500μL
Thermo Scientific™ CTC Instruments Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
Brand:  Thermo Scientific™ 365JL720
Additional Details : Weight : 0.20000kg
Product Code. 13406886
  • 2081.81 NOK / Each
In Stock
Description
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
Specifications
Specifications
22 G
Fixed Needle Syringe
0.41
500 μL