Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Brand:  Thermo Scientific™ 0012560
 View more versions of this product
Product Code. 15269633
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
  • Include flexible tubing, tubing insulation, and hose clamps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coupling
-90°C to +105°C
1 each M16 to M16 coupling
VersaCool
Coupler
M16