Learn More
Fisherbrand™ Cardboard Cryoboxes, 136mm
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryoboxes are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
350.00 NOK - 850.00 NOK
Specifications
Material | Cardboard |
---|---|
Type | Cryo Box |
Quantity | 10 Pack |
Dimensions (L x W) | 136 x 136 mm |
Length (Metric) | 136 mm |
Product Code | Brand | Color | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
11822463
|
Fisherbrand™
|
Red | 136 x 136 x 100 mm |
850.00 NOK
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
11822453
|
Fisherbrand™
|
Red | 136 x 136 x 50 mm |
398.00 NOK
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
11872453
|
Fisherbrand™
|
Red | 136 x 136 x 75 mm |
505.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11842453
|
Fisherbrand™
|
Yellow | 136 x 136 x 50 mm |
398.00 NOK
Pack of 10 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
11557633
|
Fisherbrand™
|
White | 136 x 136 x 100 mm |
685.00 NOK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11852463
|
Fisherbrand™
|
White | 136 x 136 x 130 mm |
755.00 NOK
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11862443
|
Fisherbrand™
|
Red | 136 x 136 x 32 mm |
635.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11862463
|
Fisherbrand™
|
Blue | 136 x 136 x 130 mm |
755.00 NOK
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
11852443
|
Fisherbrand™
|
Blue | 136 x 136 x 32 mm |
635.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11812463
|
Fisherbrand™
|
Blue | 136 x 136 x 100 mm |
850.00 NOK
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
11527643
|
Fisherbrand™
|
Red | 136 x 136 x 130 mm |
755.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11507623
|
Fisherbrand™
|
White | 136 x 136 x 32 mm |
350.00 NOK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11507633
|
Fisherbrand™
|
White | 136 x 136 x 75 mm |
620.00 NOK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11812453
|
Fisherbrand™
|
Blue | 136 x 136 x 50 mm |
429.00 NOK
Pack of 10 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
11812473
|
Fisherbrand™
|
Yellow | 136 x 136 x 130 mm |
720.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11547623
|
Fisherbrand™
|
Yellow | 136 x 136 x 32 mm |
655.00 NOK
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
11882453
|
Fisherbrand™
|
Green | 136 x 136 x 75 mm |
505.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11872443
|
Fisherbrand™
|
Green | 136 x 136 x 32 mm |
635.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11587623
|
Fisherbrand™
|
Green | 136 x 136 x 50 mm |
402.00 NOK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11842463
|
Fisherbrand™
|
Yellow | 136 x 136 x 100 mm |
850.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11802473
|
Fisherbrand™
|
Green | 136 x 136 x 130 mm |
755.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11832463
|
Fisherbrand™
|
Green | 136 x 136 x 100 mm |
810.00 NOK
Pack of 10 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
Description
- Available with a water-repellent finish or with a waterproof plastic coating
- Highest flexibility: Cryoboxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
- The Cryoboxes can be used for larger vessels if the grid is removed
Specifications
Cardboard | |
10 Pack | |
136 mm |
Cryo Box | |
136 x 136 mm | |
136 mm |