Learn More
Hettich™ Bucket for Swing-out Rotor no. 4294, 4176/4174
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 4255
Additional Details : Weight : 0.01000kg
Description
- Bucket
- Stainless Steel
- with Screw Cap
Specifications
Hettich Rotors, Adapters and Carriers | |
Gray | |
Stainless Steel | |
98 x 138 mm |
1000 mL | |
Bucket | |
1 |