Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
152.64 NOK - 346.62 NOK
Specifications
Description | Hollow |
---|---|
Material | Borosilicate Glass |
Product Type | Hollow Stopper |
Quantity | 10/Pk. |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10495251
|
Lenz Laborglasinstrumente™
01400010 |
Thread Size: 10/19 Cone NS |
152.64 NOK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10769121
|
Lenz Laborglasinstrumente™
01400012 |
Thread Size: 12/21 Cone NS |
157.94 NOK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15320885
|
Lenz Laborglasinstrumente™
01400019 |
Thread Size: 19/26 Cone NS |
225.78 NOK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10550052
|
Lenz Laborglasinstrumente™
01400024 |
Thread Size: 24/29 Cone NS |
257.58 NOK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10493341
|
Lenz Laborglasinstrumente™
01400029 |
Thread Size: 29/32 Cone NS |
346.62 NOK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10261731
|
Lenz Laborglasinstrumente™
01400014 |
Thread Size: 14/23 Cone NS |
171.72 NOK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN 12252, form C
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Hollow | |
Borosilicate Glass | |
Hollow Stopper | |
10/Pk. |