Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
Brand: BRAND™ 144645
262.50 NOK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.09000kg
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Hollow
- Hexagonal grip
Specifications
Ground Joint Stopper | |
1/Pk. | |
NS 45/40 |
NS 45/40 | |
Borosilicate Glass | |
Conical |