missing translation for 'onlineSavingsMsg'
Learn More

ATAGO™ Digital Hand-held PEN Refractometer: PEN-PRO

For measuring hazardous and unsanitary liquids. The Atago PEN-PRO Digital Hand-held Pen Refractometer allows you to continuously measure almost any sample in the range of 0.0 to 85.0% Brix.

Brand:  ATAGO™ 3730

Additional Details : Weight : 0.01000kg

Product Code. 11446298

  • 8395.00 NOK / Each
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For measuring hazardous and unsanitary liquids.

Order Info

Optional NIST Traceable™ Calibration Certificate and storage case available.

Specifications

Specifications

0.0 to 85.0 Brix
Water
0.1% Brix
Brix
Measuring hazardous and unsanitary liquids
0.1 lb.
0.7 in.
6.2 in.
1°C to 40°C
10°C to 40°C
PEN-PRO
±0.2% Brix
Digital
2 sec.
IP65 Water resistant (Prism head: IP67)
70 g
18 mm
160 mm
10°C to 100°C
1xAAA Alkaline Battery
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers