missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA597478
Additional Details : Weight : 0.01000kg
Description
This gene encodes a protein that is expressed in the brain and may function in neuronal migration, based on functional studies of the related astrotactin 1 gene in human and mouse. A deletion at this locus has been associated with schizophrenia. Multiple transcript variants encoding different proteins have been found for this locus.Specifications
ASTN2 | |
Polyclonal | |
Unconjugated | |
ASTN2 | |
1d8, Astn2, Astnl, astrotactin 2, astrotactin 2-like, astrotactin-2, bA264C15.1, bA67K19.1, bM243L4.1 (astrotactin 2), bM264P11.1 (astrotactin 2), bM383P24.1 (astrotactin 2), bM423P8.1 (astrotactin 2), bM452J22.1, bM452J22.1 (novel protein), bM468K2.1 (astrotactin 2), Kiaa0634 | |
Rabbit | |
Protein G | |
RUO | |
23245 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
O75129 | |
ASTN2 | |
Recombinant Human Astrotactin-2 protein (108-272AA) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |