Learn More
Analtech Glass Thin Layer Chromatography (TLC) Developing Tank
Glass Thin Layer Chromatography (TLC) Developing Tank designed for the development of TLC plates.
1835.00 NOK - 3220.00 NOK
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
11770209
|
Analtech
34105-50 |
200 x 200 mm |
3220.00 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11795348
|
Analtech
34105-00 |
100 x 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Tanks are constructed of durable glass and racks are constructed of anodized aluminum.
- Tanks accommodate standard TLC plates.
- The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development.
- Thinline™ tanks accommodate two plates and require less solvent for development.
- Latch-Lid™ units hold lids in place with stainless steel latches.
- Order rack to develop maximum number of plates in a tank.
Specifications
Glass | |
1 Ea. | |
TLC Tank |