Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
3035.25 NOK valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Universal
X-Large
Style:
W20B214
W20S213
Description
- Silicone Free
- Low Linting
Specifications
Size | Universal |
For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support and Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Style | W20S213 |
Material | Non-woven Microporous Polyethylene Laminate |
Color | White |
Seam Type | Sewn |