man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Dispensers
Bottletop Diluters and Dispensers
Bottletop Diluter and Dispenser Accessories
Eppendorf™ Adaptor
Print
Eppendorf™ Adaptor
137.00 NOK - 147.00 NOK
See promotional offers below
Specifications
Product Type
Adapter
Quantity
1 Ea.
View More Specs
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Thread Size
Price
Quantity & Availability
Product Code
Brand
Thread Size
Price
Quantity & Availability
10053231
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Eppendorf™
4960800066
38 mm
137.00 NOK
Each
View alternative products
Save Up to
»
In Stock
Log in
to see stock.
Add to basket
10691122
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Eppendorf™
4960800074
40 mm
147.00 NOK
Each
View alternative products
Save Up to
»
In Stock
Log in
Specifications
Product Type
Adapter
Quantity
1 Ea.
Documents
Product Certifications
Back to Top