Learn More
Hamilton™ 700 Series Cemented Needle Syringes
This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
562.00 NOK - 16320.00 NOK
Specifications
Material | Borosilicate Glass, Stainless Steel |
---|---|
Connection Type | Cemented |
Product Code | Brand | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
10569881
|
Hamilton™
DS80465 |
25 μL | 22 G |
16320.00 NOK
Each |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11528034
|
Hamilton™
DS80865 |
500 μL | 22 G |
16320.00 NOK
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10680812
|
Hamilton™
80350 |
10 μL | 26s G |
562.00 NOK
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- The original hand-fitted Hamilton™ syringe
- Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
- Unsurpassed syringe life
Specifications
Borosilicate Glass, Stainless Steel | |
Cemented |